siRNA Details
| Gene Symbol: Xrra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748930.3 | XP_003748978.1 | 6 | 733 | ||
| XM_006229821.2 | XP_006229883.1 | 5 | 635 | |||
| GenBank mRNA | BC158768.1 | - | 6 | 575 | 7115524 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xrra1| Gene Symbol: Xrra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748930.3 | XP_003748978.1 | 6 | 733 | ||
| XM_006229821.2 | XP_006229883.1 | 5 | 635 | |||
| GenBank mRNA | BC158768.1 | - | 6 | 575 | 7115524 | |