siRNA Details
| Gene Symbol: Olr221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000200.1 | NP_001000200.1 | 1 | 493 | ||
| Gene Symbol: Olr219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001372.1 | NP_001001372.1 | 1 | 493 | ||

Olr221
Genomic Map