siRNA Details
| Gene Symbol: Tmc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012216.1 | NP_001012216.1 | 7 | 1331 | ||
| GenBank mRNA | BC088406.1 | - | 7 | 1331 | 7112487 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmc5| Gene Symbol: Tmc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012216.1 | NP_001012216.1 | 7 | 1331 | ||
| GenBank mRNA | BC088406.1 | - | 7 | 1331 | 7112487 | |