siRNA Details
Gene Symbol: Tbc1d10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108921.1 | NP_001102391.1 | 1 | 885 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
