siRNA Details
Gene Symbol: Odf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108924.1 | NP_001102394.1 | 2 | 223 | ||
XM_006230563.2 | XP_006230625.1 | 2 | 653 | |||
GenBank mRNA | BC081921.1 | - | 2 | 222 |
