siRNA Details
| Gene Symbol: Odf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108924.1 | NP_001102394.1 | 7 | 869 | ||
| XM_006230563.2 | XP_006230625.1 | 7 | 1293 | |||
| GenBank mRNA | BC081921.1 | - | 5 | 1367 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Odf3| Gene Symbol: Odf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108924.1 | NP_001102394.1 | 7 | 869 | ||
| XM_006230563.2 | XP_006230625.1 | 7 | 1293 | |||
| GenBank mRNA | BC081921.1 | - | 5 | 1367 | ||