siRNA Details
| Gene Symbol: Psmd13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108925.2 | NP_001102395.1 | 4 | 247 | ||
| GenBank mRNA | BC158732.1 | - | 4 | 247 | 9092746 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmd13| Gene Symbol: Psmd13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108925.2 | NP_001102395.1 | 4 | 247 | ||
| GenBank mRNA | BC158732.1 | - | 4 | 247 | 9092746 | |