siRNA Details
| Gene Symbol: Cabp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108926.1 | NP_001102396.1 | 3 | 567 | ||
| XM_006230867.2 | XP_006230929.1 | 3 | 1872 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cabp4| Gene Symbol: Cabp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108926.1 | NP_001102396.1 | 3 | 567 | ||
| XM_006230867.2 | XP_006230929.1 | 3 | 1872 | |||