siRNA Details
| Gene Symbol: Slc22a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034943.1 | NP_001030115.1 | 1 | 554 | ||
| XM_008760161.1 | XP_008758383.1 | 2 | 1721 | |||
| XM_008760162.1 | XP_008758384.1 | 2 | 1569 | |||
| GenBank mRNA | BC103638.1 | - | 1 | 554 | 7109619 | |

Slc22a12
Genomic Map