siRNA Details
| Gene Symbol: Fra10ac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014246.1 | NP_001014268.1 | 7 | 563 | ||
| XM_008760371.1 | XP_008758593.1 | 7 | 1552 | |||
| XM_008760372.1 | XP_008758594.1 | 7 | 1552 | |||
| GenBank mRNA | BC090036.1 | - | 7 | 563 | 7368097 | |

Fra10ac1
Genomic Map