siRNA Details
Gene Symbol: Cpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053526.2 | NP_445978.1 | 6 | 1121 | ||
GenBank mRNA | AB042599.1 | - | 6 | 1130 | ||
BC088124.1 | - | 6 | 1121 | 7326031 | ||
FQ219519.1 | - | 4 | 406 |
