siRNA Details
| Gene Symbol: Cfap43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231588.2 | XP_006231650.1 | 29 | 3558 | ||
| XM_008760497.1 | XP_008758719.1 | 32 | 4196 | |||
| XM_345041.7 | XP_345042.5 | 33 | 4346 | |||
| GenBank mRNA | BC092207.1 | - | 8 | 855 | ||

Cfap43
Genomic Map