siRNA Details
| Gene Symbol: Trim40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009175.1 | NP_001009175.1 | 1 | 190 | ||
| XM_008772696.1 | XP_008770918.1 | 1 | 190 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim40| Gene Symbol: Trim40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009175.1 | NP_001009175.1 | 1 | 190 | ||
| XM_008772696.1 | XP_008770918.1 | 1 | 190 | |||