siRNA Details
| Gene Symbol: Skiv2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034093.1 | NP_001029265.1 | 2 | 495 | ||
| GenBank mRNA | CB576727.1 | - | 2 | 495 | ||
| CB724841.1 | - | 2 | 250 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Skiv2l2| Gene Symbol: Skiv2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034093.1 | NP_001029265.1 | 2 | 495 | ||
| GenBank mRNA | CB576727.1 | - | 2 | 495 | ||
| CB724841.1 | - | 2 | 250 | |||