siRNA Details
| Gene Symbol: Paip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108937.1 | NP_001102407.1 | 3 | 402 | ||
| XM_008760778.1 | XP_008759000.1 | 3 | 700 | |||
| XM_008760779.1 | XP_008759001.1 | 3 | 459 | |||
| XM_008760780.1 | XP_008759002.1 | 3 | 681 | |||
| GenBank mRNA | BC166996.1 | - | 3 | 273 | ||

Paip1
Genomic Map