siRNA Details
| Gene Symbol: LOC365791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761013.1 | XP_008759235.1 | 2, 3 | 419 | ||
| GenBank mRNA | BC081933.1 | - | 290 | 7114567 | ||
| Gene Symbol: LOC102549888 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232319.2 | XP_006232381.2 | 2, 3 | 419 | ||

LOC365791
Genomic Map