siRNA Details
| Gene Symbol: Rab33b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108944.1 | NP_001102414.1 | 2 | 1054 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab33b| Gene Symbol: Rab33b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108944.1 | NP_001102414.1 | 2 | 1054 | ||