siRNA Details
| Gene Symbol: Ufm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126080.2 | NP_001119552.1 | 4 | 222 | ||
| GenBank mRNA | BC091395.1 | - | 4 | 158 | 7323172 | |
| CB787557.1 | - | 4 | 260 | |||
| CV112030.1 | - | 3 | 112 | |||
| FQ229705.1 | - | 4 | 193 | |||

Ufm1
Genomic Map