siRNA Details
| Gene Symbol: Fstl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271195.1 | NP_001258124.1 | 5 | 1300 | ||
| XM_008761106.1 | XP_008759328.1 | 2 | 202 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001253719, NM_178673, AK140611, AK138388, AK033652, and others.

Fstl5
Genomic Map