siRNA Details
| Gene Symbol: Clk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014254.1 | NP_001014276.1 | 8, 9 | 1208 | ||
| XM_008761185.1 | XP_008759407.1 | 8, 9 | 1413 | |||
| XM_008761186.1 | XP_008759408.1 | 9, 10 | 1280 | |||
| XM_008761187.1 | XP_008759409.1 | 8, 9 | 1410 | |||
| XM_008761188.1 | XP_008759410.1 | 8, 9 | 1392 | |||
| XM_008761189.1 | XP_008759411.1 | 8, 9 | 1389 | |||
| XM_008761190.1 | XP_008759412.1 | 8, 9 | 1389 | |||
| XM_008761191.1 | XP_008759413.1 | 8, 9 | 1386 | |||
| XM_008761192.1 | XP_008759414.1 | 8, 9 | 1259 | |||
| XM_008761193.1 | XP_008759415.1 | 5, 6 | 1060 | |||
| XM_008761194.1 | XP_008759416.1 | 5, 6 | 539 | |||
| GenBank mRNA | BC083788.1 | - | 8, 9 | 1208 | 7098251 | |

Clk2
Genomic Map