siRNA Details
| Gene Symbol: Hormad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108949.1 | NP_001102419.1 | 6, 7 | 371 | ||
| XM_006232951.2 | XP_006233013.1 | 5, 6 | 326 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hormad1| Gene Symbol: Hormad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108949.1 | NP_001102419.1 | 6, 7 | 371 | ||
| XM_006232951.2 | XP_006233013.1 | 5, 6 | 326 | |||