siRNA Details
| Gene Symbol: Ciart | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134593.1 | NP_001128065.1 | 5 | 751 | ||
| XM_008761317.1 | XP_008759539.1 | 10 | 3222 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ciart| Gene Symbol: Ciart | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134593.1 | NP_001128065.1 | 5 | 751 | ||
| XM_008761317.1 | XP_008759539.1 | 10 | 3222 | |||