siRNA Details
| Gene Symbol: Aph1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014255.1 | NP_001014277.1 | 2 | 465 | ||
| XM_006232977.2 | XP_006233039.1 | 2 | 518 | |||
| GenBank mRNA | BC087081.1 | - | 2 | 465 | 7189126 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aph1a| Gene Symbol: Aph1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014255.1 | NP_001014277.1 | 2 | 465 | ||
| XM_006232977.2 | XP_006233039.1 | 2 | 518 | |||
| GenBank mRNA | BC087081.1 | - | 2 | 465 | 7189126 | |