siRNA Details
| Gene Symbol: Hist2h2aa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_345255.7 | XP_345256.3 | 1 | 548 | ||
| Gene Symbol: LOC690131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749350.3 | XP_003749398.1 | 1 | 538 | ||

Hist2h2aa3
Genomic Map