siRNA Details
| Gene Symbol: Trim33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233100.2 | XP_006233162.1 | 7 | 1383 | ||
| XM_345266.7 | XP_345267.3 | 7 | 1383 | |||
| GenBank mRNA | FQ227983.1 | - | 5 | 536 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim33| Gene Symbol: Trim33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233100.2 | XP_006233162.1 | 7 | 1383 | ||
| XM_345266.7 | XP_345267.3 | 7 | 1383 | |||
| GenBank mRNA | FQ227983.1 | - | 5 | 536 | ||