siRNA Details
| Gene Symbol: Slc9b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173973.1 | NP_001167444.1 | 8, 9 | 1364 | ||
| XM_008761518.1 | XP_008759740.1 | 7, 8 | 880 | |||
| GenBank mRNA | BC158723.1 | - | 3, 4 | 361 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc9b1| Gene Symbol: Slc9b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173973.1 | NP_001167444.1 | 8, 9 | 1364 | ||
| XM_008761518.1 | XP_008759740.1 | 7, 8 | 880 | |||
| GenBank mRNA | BC158723.1 | - | 3, 4 | 361 | ||