siRNA Details
| Gene Symbol: Bhmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014256.2 | NP_001014278.2 | 6 | 783 | ||
| GenBank mRNA | BC079366.1 | - | 6 | 783 | 7109516 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bhmt2| Gene Symbol: Bhmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014256.2 | NP_001014278.2 | 6 | 783 | ||
| GenBank mRNA | BC079366.1 | - | 6 | 783 | 7109516 | |