siRNA Details
Gene Symbol: Bhmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014256.2 | NP_001014278.2 | 6 | 719 | ||
GenBank mRNA | BC079366.1 | - | 6 | 719 | 7109516 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bhmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014256.2 | NP_001014278.2 | 6 | 719 | ||
GenBank mRNA | BC079366.1 | - | 6 | 719 | 7109516 |