siRNA Details
| Gene Symbol: Zdhhc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013239.1 | NP_001013257.1 | 5 | 673 | ||
| GenBank mRNA | AY886528.1 | - | 5 | 673 | ||
| BC076393.1 | - | 5 | 673 | 7107692 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zdhhc12| Gene Symbol: Zdhhc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013239.1 | NP_001013257.1 | 5 | 673 | ||
| GenBank mRNA | AY886528.1 | - | 5 | 673 | ||
| BC076393.1 | - | 5 | 673 | 7107692 | ||