siRNA Details
| Gene Symbol: Galnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015032.3 | NP_001015032.2 | 6 | 1488 | ||
| XM_006234290.2 | XP_006234352.1 | 7 | 1723 | |||
| GenBank mRNA | AB040674.1 | - | 6 | 1377 | ||
| BC101886.1 | - | 6 | 1491 | 7929522 | ||

Galnt3
Genomic Map