siRNA Details
| Gene Symbol: B3galt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108954.1 | NP_001102424.1 | 2 | 837 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B3galt1| Gene Symbol: B3galt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108954.1 | NP_001102424.1 | 2 | 837 | ||