siRNA Details
Gene Symbol: PSTPIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321135.1 | NP_001308064.1 | 6 | 886 | ||
NM_001321136.1 | NP_001308065.1 | 7 | 1105 | |||
NM_001321137.1 | NP_001308066.1 | 7 | 702 | |||
NM_003978.4 | NP_003969.2 | 6 | 886 | |||
NR_135552.1 | 6 | 886 | ||||
XM_006720737.2 | XP_006720800.1 | 6 | 618 | |||
XM_011522163.1 | XP_011520465.1 | 7 | 904 | |||
XM_011522164.1 | XP_011520466.1 | 7 | 572 | |||
XM_011522165.2 | XP_011520467.1 | 6 | 427 | |||
XM_011522166.1 | XP_011520468.1 | 7 | 904 | |||
XM_011522167.1 | XP_011520469.1 | 7 | 904 | |||
XM_011522168.2 | XP_011520470.1 | 7 | 904 | |||
XM_011522169.1 | XP_011520471.1 | 7 | 904 | |||
GenBank mRNA | AB451310.1 | - | 6 | 397 | ||
AB451440.1 | - | 6 | 397 | |||
AF038602.1 | - | 6 | 397 | |||
AF038603.1 | - | 6 | 397 | |||
AK298733.1 | - | 6 | 639 | |||
AK304052.1 | - | 6 | 675 | |||
AK308462.1 | - | 7 | 702 | |||
AK309856.1 | - | 7 | 1068 | |||
AK310072.1 | - | 7 | 819 | |||
AK316220.1 | - | 6 | 649 | |||
BC008602.1 | - | 6 | 758 | 4180398 | ||
KU178458.1 | - | 6 | 397 | |||
KU178459.1 | - | 6 | 397 | |||
U94778.1 | - | 6 | 613 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
