siRNA Details
| Gene Symbol: Olr745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000578.1 | NP_001000578.1 | 1 | 593 | ||
| Gene Symbol: Olr736 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001384.1 | NP_001001384.1 | 1 | 593 | ||

Olr745
Genomic Map