siRNA Details
| Gene Symbol: Spi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005892.2 | NP_001005892.1 | 4 | 595 | ||
| XM_006234532.2 | XP_006234594.1 | 4 | 1474 | |||
| GenBank mRNA | AB154364.1 | - | 4 | 458 | ||
| BC091185.1 | - | 4 | 595 | 7366435 | ||

Spi1
Genomic Map