siRNA Details
| Gene Symbol: Olr777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000579.1 | NP_001000579.1 | 1 | 130 | ||
| Gene Symbol: Olr774 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000918.1 | NP_001000918.1 | 1 | 130 | ||
| Gene Symbol: Olr776 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000608.1 | NP_001000608.1 | 1 | 130 | ||

Olr777
Genomic Map