siRNA Details
| Gene Symbol: Olr791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000580.1 | NP_001000580.1 | 1 | 673 | ||
| Gene Symbol: Olr788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000602.1 | NP_001000602.1 | 1 | 673 | ||

Olr791
Genomic Map