siRNA Details
| Gene Symbol: Tmco5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762110.1 | XP_008760332.1 | 7 | 540 | ||
| XM_575205.5 | XP_575205.2 | 7 | 484 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmco5b| Gene Symbol: Tmco5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762110.1 | XP_008760332.1 | 7 | 540 | ||
| XM_575205.5 | XP_575205.2 | 7 | 484 | |||