siRNA Details
| Gene Symbol: Catsper2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012220.1 | NP_001012220.1 | 3 | 505 | ||
| XM_006234860.2 | XP_006234922.1 | 3 | 551 | |||
| XM_008762187.1 | XP_008760409.1 | 3 | 187 | |||
| GenBank mRNA | BC079422.1 | - | 3 | 505 | 7114001 | |

Catsper2
Genomic Map