siRNA Details
| Gene Symbol: Apmap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034003.1 | NP_001029175.1 | 7, 8 | 990 | ||
| XM_006235215.2 | XP_006235277.1 | 8, 9 | 1124 | |||
| XM_006235217.2 | XP_006235279.1 | 8, 9 | 1007 | |||
| XM_008762362.1 | XP_008760584.1 | 8, 9 | 1538 | |||
| GenBank mRNA | BC090021.1 | - | 3, 4 | 474 | ||
| BC105824.1 | - | 7, 8 | 990 | 7372968 | ||

Apmap
Genomic Map