siRNA Details
| Gene Symbol: Rab22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108966.1 | NP_001102436.1 | 2 | 331 | ||
| XM_006235705.2 | XP_006235767.1 | 2 | 486 | |||
| XM_008762529.1 | XP_008760751.1 | 2 | 486 | |||
| GenBank mRNA | BC158650.1 | - | 2 | 310 | 8367507 | |

Rab22a
Genomic Map