siRNA Details
| Gene Symbol: Edn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077650.1 | NP_001071118.1 | 3, 4 | 565 | ||
| GenBank mRNA | S39779.1 | - | 3, 4 | 565 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Edn3| Gene Symbol: Edn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077650.1 | NP_001071118.1 | 3, 4 | 565 | ||
| GenBank mRNA | S39779.1 | - | 3, 4 | 565 | ||