siRNA Details
Gene Symbol: Edn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077650.1 | NP_001071118.1 | 2 | 337 | ||
GenBank mRNA | S39779.1 | - | 2 | 337 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Edn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077650.1 | NP_001071118.1 | 2 | 337 | ||
GenBank mRNA | S39779.1 | - | 2 | 337 |