siRNA Details
Gene Symbol: Prpf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079766.1 | NP_001073234.1 | 7 | 933 | ||
GenBank mRNA | BC128779.1 | - | 7 | 933 | 7191091 | |
FQ221925.1 | - | 7 | 936 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prpf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079766.1 | NP_001073234.1 | 7 | 933 | ||
GenBank mRNA | BC128779.1 | - | 7 | 933 | 7191091 | |
FQ221925.1 | - | 7 | 936 |