siRNA Details
Gene Symbol: RGD1310641 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079701.1 | NP_001073169.1 | 10 | 1335 | ||
XM_006237999.2 | XP_006238061.1 | 10 | 1392 | |||
XM_008763628.1 | XP_008761850.1 | 10 | 1248 | |||
XM_008763629.1 | XP_008761851.1 | 9 | 1233 | |||
GenBank mRNA | BC098003.1 | - | 3 | 269 | ||
BC128759.1 | - | 10 | 1335 | 7112639 |
