siRNA Details
| Gene Symbol: Cdc26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013240.2 | NP_001013258.1 | 4 | 444 | ||
| NM_001267054.1 | NP_001253983.1 | 4 | 372 | |||
| NM_001267055.1 | NP_001253984.1 | 3 | 336 | |||
| XM_006238241.2 | XP_006238303.1 | 4 | 460 | |||
| XM_006238242.2 | XP_006238304.1 | 3 | 348 | |||
| XM_008763796.1 | XP_008762018.1 | 4 | 383 | |||
| GenBank mRNA | AY156922.1 | - | 4 | 440 | ||
| BC086343.1 | - | 4 | 433 | 7301739 | ||
| DY317988.1 | - | 3 | 331 | |||
| FM103539.1 | - | 3 | 272 | |||
| FQ217142.1 | - | 3 | 330 | |||

Cdc26
Genomic Map