siRNA Details
Gene Symbol: Btf3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_345561.7 | XP_345562.4 | 1, 2 | 111 | ||
GenBank mRNA | FQ222777.1 | - | 1, 2 | 108 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Btf3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_345561.7 | XP_345562.4 | 1, 2 | 111 | ||
GenBank mRNA | FQ222777.1 | - | 1, 2 | 108 |