siRNA Details
| Gene Symbol: Ptch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108975.1 | NP_001102445.1 | 3 | 356 | ||
| XM_008764050.1 | XP_008762272.1 | 2 | 1045 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptch2| Gene Symbol: Ptch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108975.1 | NP_001102445.1 | 3 | 356 | ||
| XM_008764050.1 | XP_008762272.1 | 2 | 1045 | |||