siRNA Details
| Gene Symbol: Olr857 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000583.1 | NP_001000583.1 | 1 | 148 | ||
| Gene Symbol: LOC100912451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764124.1 | XP_008762346.1 | 148 | |||

Olr857
Genomic Map