siRNA Details
| Gene Symbol: Olr859 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000585.1 | NP_001000585.1 | 1 | 351 | ||
| Gene Symbol: LOC100910682 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750017.2 | XP_003750065.1 | 351 | |||

Olr859
Genomic Map