siRNA Details
| Gene Symbol: Pigv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010966.1 | NP_001010966.1 | 3 | 620 | ||
| XM_006239051.2 | XP_006239113.1 | 1 | 1385 | |||
| XM_006239055.2 | XP_006239117.1 | 3 | 366 | |||
| XM_008764159.1 | XP_008762381.1 | 3 | 1799 | |||
| XM_008764160.1 | XP_008762382.1 | 3 | 343 | |||
| GenBank mRNA | AB196341.1 | - | 3 | 620 | ||

Pigv
Genomic Map